Assay Details
Target Gene Details
Entrez Gene ID: | 157570 |
Gene Name: | establishment of sister chromatid cohesion N-acetyltransferase 2 |
Gene Aliases: |
2410004I17Rik, EFO2, RBS |
Location: |
Chr.8:27771949-27812404 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESCO2 | NM_001017420.2 | NP_001017420.1 | ||
| XM_011544421.2 | XP_011542723.1 | |||
| XM_011544422.2 | XP_011542724.1 | |||
| AF306675.1 | ||||
| AF306677.1 | 1 | 180 | ||
| AF306678.1 | ||||
| AF306679.1 | ||||
| AK124215.1 | ||||
| AL832666.1 | ||||
| AY882862.1 | AAX68677.1 | |||
| BC034641.1 | AAH34641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017032 | Chr.8:27767011 - 27821659 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map