Assay Details
Target Gene Details
Entrez Gene ID: | 378884 |
Gene Name: | NHL repeat containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
EPM2A, EPM2B, MALIN, bA204B7.2 |
Location: |
Chr.6:18120487-18122620 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NHLRC1 | NM_198586.2 | 1 | 2051 | NP_940988.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv601049 | Chr.6:18099363 - 18131233 on Build GRCh38 | Gain |
|
| nsv1034755 | Chr.6:18064320 - 18173781 on Build GRCh38 | Loss |
|
| nsv830597 | Chr.6:17953708 - 18123601 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map