Assay Details
Target Gene Details
Entrez Gene ID: | 5569 |
Gene Name: | protein kinase (cAMP-dependent, catalytic) inhibitor alpha |
Gene Aliases: |
PRKACN1 |
Location: |
Chr.8:78516101-78605267 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKIA | NM_006823.3 | NP_006814.1 | ||
| NM_181839.2 | NP_862822.1 | |||
| S76965.1 | AAB21141.1 |
More Information
Set Membership: |
|

Genomic Map