Assay Details
Target Gene Details
Entrez Gene ID: | 60528 |
Gene Name: | elaC ribonuclease Z 2 |
Gene Aliases: |
COXPD17, ELC2, HPC2 |
Location: |
Chr.17:12991612-13018064 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELAC2 | NM_001165962.1 | 23 | 2718 | NP_001159434.1 |
| NM_018127.6 | 24 | 2838 | NP_060597.4 | |
| NM_173717.1 | 24 | 2835 | NP_776065.1 | |
| AF304370.1 | 24 | 2721 | AAG24441.1 | |
| AK001392.1 | 24 | 2792 | ||
| AK074244.1 | 16 | 1960 | ||
| AK094012.1 | 9 | 2608 | ||
| AK094333.1 | 16 | 2549 | ||
| AK094687.1 | 24 | 2722 | ||
| AK124838.1 | 13 | 2267 | BAC85964.1 | |
| AK125030.1 | 22 | 3551 | BAC86026.1 | |
| AK298397.1 | ||||
| AK300011.1 | ||||
| BC001939.1 | 24 | 2781 | AAH01939.1 | |
| BC004158.2 | 24 | 2781 | AAH04158.1 | |
| CB241821.1 | 1 | 211 |
More Information
Set Membership: |
|

Genomic Map