Assay Details
Target Gene Details
Entrez Gene ID: | 192669 |
Gene Name: | argonaute 3, RISC catalytic component |
Gene Aliases: |
EIF2C3 |
Location: |
Chr.1:35925198-36072037 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO3 | NM_024852.3 | 2 | 415 | NP_079128.2 |
| NM_177422.2 | 2 | 415 | NP_803171.1 | |
| XM_005270575.3 | 3 | 5869 | XP_005270632.1 | |
| XM_011540879.2 | 2 | 1297 | XP_011539181.1 | |
| XM_017000523.1 | 3 | 5885 | XP_016856012.1 | |
| AK022827.1 | 2 | 170 | BAB14262.1 | |
| AK027796.1 | 3 | 349 | BAB55374.1 | |
| AK292870.1 | 2 | 381 | ||
| BC066888.1 | 2 | 415 | AAH66888.1 | |
| BF438817.1 | 2 | 326 | ||
| BF979526.1 | 2 | 420 |
More Information
Set Membership: |
|

Genomic Map