Assay Details
Target Gene Details
Entrez Gene ID: | 64689 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
GOLPH5, GRASP65, P65 |
Location: |
Chr.3:39096599-39108363 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GORASP1 | NM_001278789.1 | 1 | 264 | NP_001265718.1 |
| NM_001278790.1 | 1 | 264 | NP_001265719.1 | |
| NM_031899.3 | 1 | 727 | NP_114105.1 | |
| NR_103866.1 | 1 | 264 | ||
| NR_103867.1 | 1 | 264 | ||
| XM_017007050.1 | XP_016862539.1 | |||
| XM_017007051.1 | 1 | 442 | XP_016862540.1 | |
| XM_017007052.1 | XP_016862541.1 | |||
| AJ409349.1 | CAC35160.1 | |||
| AK001574.1 | ||||
| AK057043.1 | ||||
| AK124755.1 | 1 | 727 | ||
| AK300271.1 | ||||
| AK303847.1 | ||||
| BC008928.2 | AAH08928.1 | |||
| BC075854.1 | AAH75854.1 | |||
| BC103826.1 | AAI03827.1 | |||
| BC103827.1 | AAI03828.1 | |||
| DB031221.1 |
Target Gene Details
Entrez Gene ID: | 199223 |
Gene Name: | tetratricopeptide repeat domain 21A |
Gene Aliases: |
IFT139A, STI2 |
Location: |
Chr.3:39107661-39139009 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC21A | NM_001105513.2 | NP_001098983.2 | ||
| NM_145755.2 | NP_665698.2 | |||
| DC403032.1 |
More Information
Set Membership: |
|

Genomic Map