Assay Details
Target Gene Details
Entrez Gene ID: | 54714 |
Gene Name: | cyclic nucleotide gated channel beta 3 |
Gene Aliases: |
ACHM1 |
Location: |
Chr.8:86573608-86743675 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNGB3 | NM_019098.4 | 18 | 3511 | NP_061971.3 |
| XM_011517138.2 | 16 | 3166 | XP_011515440.1 | |
| AF272900.1 | 18 | 3509 | AAF86274.1 | |
| BX118844.1 | 1 | 394 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422239 | Chr.8:86535165 - 87814698 on Build GRCh38 | Duplication |
|
| nsv1023510 | Chr.8:86416772 - 86862720 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map