Assay Details
Target Gene Details
Entrez Gene ID: | 1718 |
Gene Name: | 24-dehydrocholesterol reductase |
Gene Aliases: |
DCE, Nbla03646, SELADIN1, seladin-1 |
Location: |
Chr.1:54849627-54887248 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHCR24 | NM_014762.3 | 9 | 1728 | NP_055577.1 |
| AB073391.1 | 10 | 1603 | BAE45721.1 | |
| AF261758.1 | 9 | 1698 | AAG17288.1 | |
| AK298414.1 | 4 | 957 | ||
| AK302774.1 | 9 | 1559 | ||
| AK304302.1 | 6 | 1016 | ||
| AK304421.1 | 5 | 855 | ||
| AK304437.1 | 9 | 1445 | ||
| AK316419.1 | 9 | 1490 | ||
| AK316427.1 | 9 | 1429 | ||
| BC004375.1 | 9 | 1634 | AAH04375.1 | |
| BC011669.2 | 9 | 1661 | AAH11669.1 | |
| D13643.3 | 9 | 1711 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461561 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
| nsv829915 | Chr.1:54828940 - 54992690 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map