Assay Details
Target Gene Details
Entrez Gene ID: | 7403 |
Gene Name: | lysine demethylase 6A |
Gene Aliases: |
KABUK2, UTX, bA386N14.2 |
Location: |
Chr.X:44873175-45112612 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
More Information
Set Membership: |
|

Genomic Map