Assay Details
Target Gene Details
Entrez Gene ID: | 5018 |
Gene Name: | OXA1L, mitochondrial inner membrane protein |
Gene Aliases: |
OXA1 |
Location: |
Chr.14:22766522-22771789 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXA1L | NM_005015.3 | NP_005006.3 | ||
| AK298249.1 | ||||
| BC001669.2 | AAH01669.2 | |||
| BC112331.1 | AAI12332.1 | |||
| BX248001.1 | CAD62333.1 | |||
| BX248295.1 | CAD62623.1 | |||
| X80695.1 | CAA56712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv127n21 | Chr.14:22690985 - 22790490 on Build GRCh38 | Gain |
|
| nsv563998 | Chr.14:22414183 - 22879743 on Build GRCh38 | Gain |
|
| dgv1836n100 | Chr.14:22676576 - 22803700 on Build GRCh38 | Gain |
|
| esv3633763 | Chr.14:22686795 - 22843098 on Build GRCh38 | Gain |
|
| nsv564039 | Chr.14:22629313 - 22794929 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map