Assay Details
Target Gene Details
Entrez Gene ID: | 221458 |
Gene Name: | kinesin family member 6 |
Gene Aliases: |
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1 |
Location: |
Chr.6:39335100-39725405 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF6 | NM_001289020.1 | 22 | 2771 | NP_001275949.1 |
| NM_001289021.1 | 22 | 2654 | NP_001275950.1 | |
| NM_001289024.1 | 10 | 1295 | NP_001275953.1 | |
| NM_145027.4 | 23 | 2822 | NP_659464.3 | |
| XM_011514359.2 | 22 | 2696 | XP_011512661.1 | |
| AK123217.1 | 10 | 1219 | BAC85560.1 | |
| AK131471.1 | 23 | 2822 | BAD18616.1 | |
| AL832634.2 | 10 | 1058 | CAD89950.1 | |
| BC022074.2 | 10 | 1236 | AAH22074.1 | |
| BX649045.1 | 19 | 2351 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830645 | Chr.6:39259825 - 39440534 on Build GRCh38 | Loss |
|
| nsv830644 | Chr.6:39148754 - 39358204 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74659777] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map