Assay Details
Target Gene Details
Entrez Gene ID: | 120379 |
Gene Name: | PIH1 domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.11:112052161-112074288 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIH1D2 | NM_001082619.1 | NP_001076088.1 | ||
| NM_138789.3 | NP_620144.1 | |||
| XM_011542593.2 | XP_011540895.2 | |||
| XM_017017201.1 | XP_016872690.1 | |||
| XM_017017202.1 | XP_016872691.1 | |||
| XM_017017203.1 | XP_016872692.1 | |||
| XM_017017204.1 | XP_016872693.1 | |||
| XM_017017205.1 | XP_016872694.1 | |||
| AK300496.1 | ||||
| BC019238.2 | AAH19238.1 | |||
| CB159963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map