Assay Details
Target Gene Details
Entrez Gene ID: | 3578 |
Gene Name: | interleukin 9 |
Gene Aliases: |
HP40, IL-9, P40 |
Location: |
Chr.5:135892246-135895827 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL9 | NM_000590.1 | NP_000581.1 | ||
| BC066283.1 | AAH66283.1 | |||
| BC066284.1 | AAH66284.1 | |||
| BC066285.1 | AAH66285.1 | |||
| BC066286.1 | AAH66286.1 | |||
| BC066287.1 | AAH66287.1 | |||
| M30134.1 | AAA36400.1 | |||
| S63356.1 | AAB19823.1 | |||
| X17543.1 | CAA35581.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759380 | Chr.5:135829912 - 136139310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map