Assay Details
Target Gene Details
Entrez Gene ID: | 4296 |
Gene Name: | mitogen-activated protein kinase kinase kinase 11 |
Gene Aliases: |
MEKK11, MLK-3, MLK3, PTK1, SPRK |
Location: |
Chr.11:65597755-65614338 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K11 | NM_002419.3 | 1 | 534 | NP_002410.1 |
| AB209655.1 | 1 | 521 | BAD92892.1 | |
| AK092015.1 | 1 | 1667 | ||
| AK222781.1 | 1 | 527 | BAD96501.1 | |
| AK315557.1 | 1 | 506 | ||
| AU119082.2 | 1 | 534 | ||
| BC011263.2 | 1 | 522 | AAH11263.1 | |
| BC064543.1 | 1 | 493 | AAH64543.1 | |
| L32976.1 | 1 | 522 | AAA59859.1 | |
| U07747.1 | 1 | 491 | AAA19647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1969n54 | Chr.11:65568234 - 65663889 on Build GRCh38 | Loss |
|
| esv2761680 | Chr.11:65576857 - 65623846 on Build GRCh38 | Gain |
|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map