Assay Details
Target Gene Details
Entrez Gene ID: | 8187 |
Gene Name: | zinc finger protein 239 |
Gene Aliases: |
HOK-2, MOK2 |
Location: |
Chr.10:43554516-43574619 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF239 | NM_001099282.1 | 4 | 326 | NP_001092752.1 |
| NM_001099283.1 | 2 | 163 | NP_001092753.1 | |
| NM_001099284.1 | 3 | 286 | NP_001092754.1 | |
| NM_001324347.1 | 3 | 204 | NP_001311276.1 | |
| NM_001324348.1 | 2 | 232 | NP_001311277.1 | |
| NM_001324349.1 | 4 | 396 | NP_001311278.1 | |
| NM_001324350.1 | 3 | 273 | NP_001311279.1 | |
| NM_001324351.1 | 3 | 355 | NP_001311280.1 | |
| NM_001324352.1 | 4 | 331 | NP_001311281.1 | |
| NM_001324353.1 | 5 | 436 | NP_001311282.1 | |
| NM_005674.2 | 2 | 645 | NP_005665.2 | |
| XM_005271832.2 | 2 | 158 | XP_005271889.1 | |
| XM_006718001.2 | 2 | 257 | XP_006718064.1 | |
| XM_006718003.3 | 2 | 157 | XP_006718066.1 | |
| XM_011540232.2 | 3 | 3244 | XP_011538534.1 | |
| XM_011540234.2 | 3 | 291 | XP_011538536.1 | |
| XM_011540235.2 | 3 | 290 | XP_011538537.1 | |
| XM_011540236.2 | 3 | 299 | XP_011538538.1 | |
| XM_011540237.2 | 3 | 249 | XP_011538539.1 | |
| XM_011540238.2 | 2 | 101 | XP_011538540.1 | |
| XM_017016740.1 | 4 | 328 | XP_016872229.1 | |
| AK292013.1 | 3 | 282 | ||
| BC026030.1 | 4 | 270 | AAH26030.1 | |
| CK820210.1 | 5 | 378 | ||
| DA597035.1 | 4 | 310 | ||
| DA686973.1 | 2 | 159 | ||
| DA937379.1 | 3 | 282 | ||
| DN994387.1 | 3 | 331 | ||
| X82125.1 | 2 | 645 | CAA57637.1 |
More Information
Set Membership: |
|

Genomic Map