Assay Details
Target Gene Details
Entrez Gene ID: | 5893 |
Gene Name: | RAD52 homolog, DNA repair protein |
Gene Aliases: |
- |
Location: |
Chr.12:911028-991195 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD52 | NM_001297419.1 | 12 | 2790 | NP_001284348.1 |
| NM_001297421.1 | 10 | 2474 | NP_001284350.1 | |
| NM_134424.3 | 12 | 2678 | NP_602296.2 | |
| NR_123713.1 | 14 | 2960 | ||
| XM_005253720.4 | 13 | 3552 | XP_005253777.1 | |
| XM_005253721.2 | 12 | 2778 | XP_005253778.1 | |
| XM_011520990.2 | 13 | 2814 | XP_011519292.1 | |
| XM_011520991.2 | 12 | 2680 | XP_011519293.1 | |
| XM_011520995.2 | 8 | 2262 | XP_011519297.1 | |
| XM_017019769.1 | 13 | 2935 | XP_016875258.1 | |
| XM_017019770.1 | 11 | 2527 | XP_016875259.1 | |
| AA402047.1 | ||||
| U12134.1 | AAA85793.1 | |||
| U27516.1 | AAA87554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain |
|
| esv3628261 | Chr.12:880274 - 938296 on Build GRCh38 | Gain |
|
| nsv1042333 | Chr.12:798809 - 972343 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map