Assay Details
Target Gene Details
Entrez Gene ID: | 55341 |
Gene Name: | large 60S subunit nuclear export GTPase 1 |
Gene Aliases: |
- |
Location: |
Chr.3:194640788-194672477 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSG1 | NM_018385.2 | NP_060855.2 | ||
| XM_017006796.1 | XP_016862285.1 | |||
| AK002163.1 | BAA92116.1 | |||
| AK225094.1 | ||||
| AL136897.1 | CAB66831.1 | |||
| BC015042.1 | ||||
| BC040119.1 | ||||
| BC068500.1 | AAH68500.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599208 | Chr.3:194592316 - 194799085 on Build GRCh38 | Loss |
|
| nsv508268 | Chr.3:194618407 - 194681984 on Build GRCh38 | Deletion |
|
| nsv519476 | Chr.3:194645278 - 194972500 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map