Assay Details
Target Gene Details
Entrez Gene ID: | 3320 |
Gene Name: | heat shock protein 90 alpha family class A member 1 |
Gene Aliases: |
EL52, HEL-S-65p, HSP86, HSP89A, HSP90A, HSP90N, HSPC1, HSPCA, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP-2, LAP2 |
Location: |
Chr.14:102080738-102139749 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSP90AA1 | NM_001017963.2 | 1 | 100 | NP_001017963.2 |
| XM_011536718.2 | 1 | 100 | XP_011535020.1 | |
| AJ890082.1 | 1 | 37 | CAI64495.1 | |
| AK056446.1 | 1 | 37 | ||
| BX248761.1 | 1 | 36 | CAD66568.1 | |
| DA237149.1 | 1 | 100 |
Target Gene Details
Entrez Gene ID: | 91833 |
Gene Name: | WD repeat domain 20 |
Gene Aliases: |
DMR |
Location: |
Chr.14:102139518-102224686 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR20 | XM_017021777.1 | XP_016877266.1 | ||
| XM_017021780.1 | 1 | 133 | XP_016877269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565947 | Chr.14:102001510 - 102204240 on Build GRCh38 | Gain |
|
| dgv25n68 | Chr.14:102000993 - 102195522 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map