Assay Details
Target Gene Details
Entrez Gene ID: | 79643 |
Gene Name: | charged multivesicular body protein 6 |
Gene Aliases: |
VPS20 |
Location: |
Chr.17:80991841-81000133 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP6 | NM_024591.4 | 8 | 679 | NP_078867.2 |
| AK021811.1 | 8 | 617 | BAB13901.1 | |
| AK223187.1 | 8 | 641 | BAD96907.1 | |
| AK292105.1 | 8 | 645 | ||
| AY329087.1 | AAQ91196.1 | |||
| BC010108.2 | 8 | 625 | AAH10108.1 | |
| CR457284.1 | CAG33565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952386 | Chr.17:80872501 - 81005700 on Build GRCh38 | Deletion |
|
| nsv828128 | Chr.17:80659041 - 81736487 on Build GRCh38 | Loss |
|
| esv3641388 | Chr.17:80978629 - 81012536 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map