Assay Details
Target Gene Details
Entrez Gene ID: | 54585 |
Gene Name: | leucine zipper transcription factor like 1 |
Gene Aliases: |
BBS17 |
Location: |
Chr.3:45823316-45916042 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LZTFL1 | NM_001276378.1 | 12 | 2814 | NP_001263307.1 |
| NM_001276379.1 | 10 | 2585 | NP_001263308.1 | |
| NM_020347.3 | 10 | 2533 | NP_065080.1 | |
| NR_075080.1 | 9 | 2408 | ||
| XM_006713207.2 | 9 | 2414 | XP_006713270.2 | |
| XM_011533838.2 | 10 | 2457 | XP_011532140.1 | |
| XM_017006644.1 | 13 | 3171 | XP_016862133.1 | |
| XM_017006645.1 | 12 | 3046 | XP_016862134.1 | |
| AJ297351.1 | 10 | 2490 | CAB95836.1 | |
| AK027165.1 | 1 | 1161 | ||
| AK129805.1 | 1 | 577 | ||
| BC042483.1 | 10 | 2493 | AAH42483.1 | |
| BX640604.1 | 10 | 2518 | CAE45710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834678 | Chr.3:45762802 - 45968764 on Build GRCh38 | Gain |
|
| nsv3796 | Chr.3:45809469 - 45844553 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map