Assay Details
Target Gene Details
Entrez Gene ID: | 4704 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A9 |
Gene Aliases: |
CC6, CI-39k, CI39k, NDUFS2L, SDR22E1 |
Location: |
Chr.12:4649098-4687554 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA9 | NM_005002.4 | NP_004993.1 | ||
| AF050641.1 | AAD42055.1 | |||
| AK291067.1 | ||||
| AK308588.1 | ||||
| BC003351.1 | AAH03351.1 | |||
| BC009311.2 | AAH09311.1 | |||
| BC015837.1 | AAH15837.1 | |||
| BC111546.1 | AAI11547.1 | |||
| L04490.1 | AAA36350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| nsv587 | Chr.12:4636246 - 4670472 on Build GRCh38 | Insertion |
|
| nsv1036308 | Chr.12:4639491 - 4694990 on Build GRCh38 | Loss |
|
| nsv1040098 | Chr.12:3987637 - 5646161 on Build GRCh38 | Gain |
|
| nsv832318 | Chr.12:4546633 - 4766671 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map