Assay Details
Target Gene Details
Entrez Gene ID: | 51192 |
Gene Name: | chemokine-like factor |
Gene Aliases: |
C32, CKLF1, CKLF2, CKLF3, CKLF4, HSPC224, UCK-1 |
Location: |
Chr.16:66552563-66566287 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CKLF | NM_001040138.2 | 4 | 527 | NP_001035228.1 |
| NM_016326.3 | 2 | 469 | NP_057410.1 | |
| NM_016951.3 | 4 | 724 | NP_058647.1 | |
| NM_181640.2 | 3 | 565 | NP_857591.1 | |
| NM_181641.2 | 3 | 628 | NP_857592.1 | |
| AA234074.1 | ||||
| AK290307.1 | 3 | 539 | ||
| AK301240.1 | ||||
| BU189908.1 | 3 | 525 | ||
| DA113473.1 | 3 | 419 | ||
| DA804889.1 | 4 | 508 |
Target Gene Details
Entrez Gene ID: | 100529251 |
Gene Name: | CKLF-CMTM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:66552563-66579135 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CKLF-CMTM1 | NM_001202509.1 | NP_001189438.1 | ||
| NM_001204098.1 | NP_001191027.1 | |||
| NM_001204099.1 | NP_001191028.1 | |||
| BM929895.1 | ||||
| FY211334.1 | ||||
| FY212695.1 |
More Information
Set Membership: |
|

Genomic Map