Assay Details
Target Gene Details
Entrez Gene ID: | 2786 |
Gene Name: | G protein subunit gamma 4 |
Gene Aliases: |
- |
Location: |
Chr.1:235547685-235650754 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG4 | NM_001098721.1 | 4 | 636 | NP_001092191.1 |
| NM_001098722.1 | 4 | 762 | NP_001092192.1 | |
| NM_004485.3 | 3 | 524 | NP_004476.1 | |
| XM_006711761.2 | 3 | 407 | XP_006711824.1 | |
| XM_011544167.2 | 3 | 337 | XP_011542469.1 | |
| BC022485.1 | 3 | 524 | AAH22485.1 | |
| DA122711.1 | ||||
| DA230013.1 | 4 | 495 | ||
| U31382.1 | 3 | 406 | AAC50204.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv30n21 | Chr.1:235198002 - 235605078 on Build GRCh38 | Gain |
|
| nsv1013571 | Chr.1:235210731 - 235587805 on Build GRCh38 | Gain |
|
| nsv1011288 | Chr.1:235376656 - 235666302 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map