Assay Details
Target Gene Details
Entrez Gene ID: | 23741 |
Gene Name: | EP300 interacting inhibitor of differentiation 1 |
Gene Aliases: |
C15orf3, CRI1, EID-1, IRO45620, PNAS-22, PTD014, RBP21 |
Location: |
Chr.15:48878093-48880183 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EID1 | NM_014335.2 | 1 | 604 | NP_055150.1 |
| AF092135.1 | 1 | 583 | AAD40377.1 | |
| AF109873.1 | 1 | 583 | AAG35179.1 | |
| AF274947.1 | 1 | 232 | AAK07524.1 | |
| AF349444.1 | 1 | 574 | AAK29640.1 | |
| AK098383.1 | 2 | 493 | BAC05296.1 | |
| AK315365.1 | 1 | 588 | ||
| AL109701.1 | 1 | 544 | CAB52022.1 | |
| AL357456.1 | 1 | 532 | CAB93108.1 | |
| BC114944.1 | 1 | 596 | ||
| BC114946.1 | 1 | 596 | ||
| BI550673.1 | 1 | 609 |
Target Gene Details
Entrez Gene ID: | 399694 |
Gene Name: | SHC adaptor protein 4 |
Gene Aliases: |
RaLP, SHCD |
Location: |
Chr.15:48823737-48963444 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHC4 | NM_203349.3 | NP_976224.3 | ||
| XM_005254375.3 | XP_005254432.1 | |||
| AK125762.1 | ||||
| AY464565.1 | AAR19363.1 | |||
| BC033907.2 | AAH33907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833001 | Chr.15:48751446 - 48912568 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map