Assay Details
Target Gene Details
Entrez Gene ID: | 10840 |
Gene Name: | aldehyde dehydrogenase 1 family member L1 |
Gene Aliases: |
10-FTHFDH, 10-fTHF, FDH, FTHFD |
Location: |
Chr.3:126103561-126181526 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1L1 | NM_001270364.1 | NP_001257293.1 | ||
| NM_001270365.1 | NP_001257294.1 | |||
| NM_012190.3 | NP_036322.2 | |||
| NR_072979.1 | ||||
| XM_006713481.2 | XP_006713544.1 | |||
| XM_011512355.1 | XP_011510657.1 | |||
| XM_017005613.1 | XP_016861102.1 | |||
| XM_017005614.1 | XP_016861103.1 | |||
| AA639766.1 | ||||
| AB209299.1 | BAD92536.1 | |||
| AF052732.1 | AAC35000.1 | |||
| AK124908.1 | 6 | 3393 | BAC85992.1 | |
| AK222535.1 | BAD96255.1 | |||
| AK222694.1 | BAD96414.1 | |||
| AK294392.1 | ||||
| BC027241.1 | AAH27241.1 | |||
| CR749807.1 | CAH18667.1 |
Target Gene Details
Entrez Gene ID: | 100874204 |
Gene Name: | ALDH1L1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:126103640-126108069 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1L1-AS1 | NR_046602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524971 | Chr.3:126024828 - 126155545 on Build GRCh38 | Loss |
|
| nsv591465 | Chr.3:125283331 - 126140524 on Build GRCh38 | Gain |
|
| nsv955334 | Chr.3:126101258 - 126137757 on Build GRCh38 | Deletion |
|
| nsv829716 | Chr.3:125928159 - 126116976 on Build GRCh38 | Gain+Loss |
|
| nsv1011457 | Chr.3:125287676 - 126146923 on Build GRCh38 | Gain |
|
| dgv8545n54 | Chr.3:126091128 - 126251891 on Build GRCh38 | Loss |
|
| nsv520037 | Chr.3:126103429 - 126135566 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map