Assay Details
Target Gene Details
Entrez Gene ID: | 7050 |
Gene Name: | TGFB induced factor homeobox 1 |
Gene Aliases: |
HPE4, TGIF |
Location: |
Chr.18:3411927-3458412 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGIF1 | NM_001278686.1 | NP_001265615.1 | ||
| NM_173207.2 | 1 | 128 | NP_775299.1 | |
| NM_174886.2 | NP_777480.1 | |||
| AK291112.1 | 1 | 84 | ||
| BE906361.1 | ||||
| BM849453.1 | ||||
| BU657748.1 | 1 | 104 | ||
| CB996365.1 | ||||
| HY041619.1 | 1 | 129 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527676 | Chr.18:3445828 - 3452980 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map