Assay Details
Target Gene Details
Entrez Gene ID: | 3363 |
Gene Name: | 5-hydroxytryptamine receptor 7 |
Gene Aliases: |
5-HT7 |
Location: |
Chr.10:90740818-90857914 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR7 | NM_000872.4 | 2 | 1108 | NP_000863.1 |
| NM_019859.3 | 2 | 1108 | NP_062873.1 | |
| NM_019860.3 | 2 | 1108 | NP_062874.1 | |
| AB451482.1 | 2 | 865 | ||
| AK292606.1 | 2 | 878 | ||
| AY493988.1 | 2 | 865 | AAR87480.1 | |
| BC047526.1 | 2 | 1108 | AAH47526.1 | |
| L21195.1 | 2 | 892 | AAC37538.1 | |
| U68487.1 | 2 | 892 | AAB48393.1 | |
| U68488.1 | 2 | 892 | AAB48394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516743 | Chr.10:90323258 - 90845362 on Build GRCh38 | Gain |
|
| dgv952n100 | Chr.10:90321336 - 90845607 on Build GRCh38 | Gain |
|
| nsv1036058 | Chr.10:90711811 - 90845607 on Build GRCh38 | Gain |
|
| nsv467432 | Chr.10:90322493 - 90800151 on Build GRCh38 | Gain |
|
| esv2422237 | Chr.10:90180785 - 90787801 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map