Assay Details
Target Gene Details
Entrez Gene ID: | 5655 |
Gene Name: | kallikrein related peptidase 10 |
Gene Aliases: |
NES1, PRSSL1 |
Location: |
Chr.19:51012744-51020175 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK10 | NM_001077500.1 | 3 | 308 | NP_001070968.1 |
| NM_002776.4 | 3 | 407 | NP_002767.2 | |
| NM_145888.2 | 3 | 269 | NP_665895.1 | |
| XM_005259061.3 | 3 | 329 | XP_005259118.1 | |
| XM_005259062.3 | 3 | 293 | XP_005259119.1 | |
| XM_006723287.3 | 3 | 296 | XP_006723350.1 | |
| XM_006723289.3 | 3 | 391 | XP_006723352.1 | |
| XM_017026993.1 | 3 | 306 | XP_016882482.1 | |
| AF024605.1 | 3 | 269 | AAB81602.1 | |
| AK292365.1 | 3 | 398 | ||
| AK309456.1 | 2 | 495 | ||
| AY561635.1 | 3 | 336 | AAS66976.1 | |
| BC002710.2 | 3 | 407 | AAH02710.1 | |
| BM830577.1 | 3 | 308 | ||
| CR450327.1 | 2 | 188 | CAG29323.1 | |
| KJ010190.1 | 2 | 121 | ||
| KJ010191.1 | 2 | 121 | ||
| KJ010192.1 | ||||
| KJ010193.1 | ||||
| KJ010194.1 | ||||
| KJ010195.1 | ||||
| KJ010197.1 | 2 | 121 | ||
| KJ010198.1 | ||||
| KJ010201.1 | 1 | 122 | ||
| KJ010202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677627 | Chr.19:51015389 - 51020436 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map