Assay Details
Target Gene Details
Entrez Gene ID: | 5523 |
Gene Name: | protein phosphatase 2 regulatory subunit B''alpha |
Gene Aliases: |
PPP2R3, PR130, PR72 |
Location: |
Chr.3:135965673-136147910 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R3A | NM_001190447.1 | NP_001177376.1 | ||
| NM_002718.4 | NP_002709.2 | |||
| XM_017006787.1 | XP_016862276.1 | |||
| AK293012.1 | ||||
| AK298059.1 | ||||
| AK314243.1 | ||||
| BC015034.1 | ||||
| BC029409.1 | ||||
| BC065531.1 | AAH65531.1 | |||
| BE897340.1 | ||||
| DB223168.1 | ||||
| L07590.1 | AAB02613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829732 | Chr.3:135842864 - 136037998 on Build GRCh38 | Gain |
|
| nsv591829 | Chr.3:135388836 - 136050164 on Build GRCh38 | Gain |
|
| esv3597878 | Chr.3:135840563 - 135999462 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map