Assay Details
Target Gene Details
Entrez Gene ID: | 197358 |
Gene Name: | NLR family CARD domain containing 3 |
Gene Aliases: |
CLR16.2, NOD3 |
Location: |
Chr.16:3538272-3578373 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRC3 | NM_178844.3 | NP_849172.2 | ||
| NR_075083.2 | ||||
| XM_017023027.1 | XP_016878516.1 | |||
| XM_017023028.1 | XP_016878517.1 | |||
| XM_017023029.1 | XP_016878518.1 | |||
| XM_017023030.1 | XP_016878519.1 | |||
| XM_017023031.1 | XP_016878520.1 | |||
| XM_017023032.1 | XP_016878521.1 | |||
| XM_017023033.1 | XP_016878522.1 | |||
| XM_017023034.1 | XP_016878523.1 | |||
| XM_017023035.1 | XP_016878524.1 | |||
| XM_017023036.1 | XP_016878525.1 | |||
| XM_017023037.1 | XP_016878526.1 | |||
| XM_017023038.1 | XP_016878527.1 | |||
| XM_017023039.1 | XP_016878528.1 | |||
| AF501291.1 | AAM22459.1 | |||
| AF501292.1 | AAM22460.1 | |||
| AK074109.1 | BAB84935.1 | |||
| AK090431.1 | BAC03412.1 | |||
| AY601811.1 | AAT48367.1 | |||
| FJ889357.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833128 | Chr.16:3525122 - 3690497 on Build GRCh38 | Loss |
|
| nsv517451 | Chr.16:3549655 - 3611814 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map