Assay Details
Target Gene Details
Entrez Gene ID: | 9731 |
Gene Name: | centrosomal protein 104 |
Gene Aliases: |
CFAP256, GlyBP, JBTS25, KIAA0562, ROC22 |
Location: |
Chr.1:3812081-3857387 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP104 | NM_014704.3 | NP_055519.1 | ||
| XM_005244815.4 | XP_005244872.1 | |||
| XM_011542473.2 | XP_011540775.1 | |||
| XM_011542474.2 | XP_011540776.1 | |||
| XM_011542475.2 | XP_011540777.1 | |||
| XM_011542476.2 | XP_011540778.1 | |||
| XM_011542477.2 | XP_011540779.1 | |||
| XM_011542478.1 | XP_011540780.1 | |||
| XM_017002917.1 | XP_016858406.1 | |||
| XM_017002918.1 | XP_016858407.1 | |||
| XM_017002919.1 | XP_016858408.1 | |||
| AB011134.1 | ||||
| AK302190.1 | ||||
| BC001640.2 | AAH01640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain |
|
| esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain |
|
| nsv830425 | Chr.1:3740889 - 3841825 on Build GRCh38 | Gain |
|
| nsv509590 | Chr.1:3826457 - 3902492 on Build GRCh38 | Insertion |
|
| esv3585052 | Chr.1:3817921 - 3928224 on Build GRCh38 | Gain |
|
| nsv545130 | Chr.1:3826568 - 4283888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map