Assay Details
Target Gene Details
Entrez Gene ID: | 27044 |
Gene Name: | staphylococcal nuclease and tudor domain containing 1 |
Gene Aliases: |
TDRD11, p100 |
Location: |
Chr.7:127652119-128092607 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SND1 | NM_014390.2 | NP_055205.2 | ||
| XM_017011987.1 | XP_016867476.1 | |||
| AB209510.1 | BAD92747.1 | |||
| AK096583.1 | ||||
| AK304181.1 | ||||
| AK309014.1 | ||||
| AK312313.1 | ||||
| BC017180.2 | ||||
| BE799345.1 | ||||
| BT009785.1 | AAP88787.1 | |||
| U22055.1 | AAA80488.1 |
Target Gene Details
Entrez Gene ID: | 27099 |
Gene Name: | SND1 intronic transcript 1 |
Gene Aliases: |
C7orf54, NAG8, NSG-X |
Location: |
Chr.7:127997509-128000077 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SND1-IT1 | NR_027330.1 | 1 | 963 | |
| AF095687.1 | 1 | 670 | AAC96312.1 | |
| AF191492.1 | 1 | 963 | AAG28385.1 | |
| AK024176.1 | 1 | 875 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024743 | Chr.7:127858642 - 128029951 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map