Assay Details
Target Gene Details
Entrez Gene ID: | 9739 |
Gene Name: | SET domain containing 1A |
Gene Aliases: |
KMT2F, Set1, Set1A |
Location: |
Chr.16:30956618-30984664 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD1A | NM_014712.2 | NP_055527.1 | ||
| XM_005255723.1 | XP_005255780.1 | |||
| XM_006721106.3 | XP_006721169.1 | |||
| XM_017023909.1 | XP_016879398.1 | |||
| AB002337.2 | ||||
| BC035795.2 | AAH35795.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv35n68 | Chr.16:30833730 - 31036730 on Build GRCh38 | Loss |
|
| nsv571820 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
| nsv953802 | Chr.16:30876680 - 30999279 on Build GRCh38 | Deletion |
|
| nsv833193 | Chr.16:30949475 - 31122283 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map