Assay Details
Target Gene Details
Entrez Gene ID: | 3696 |
Gene Name: | integrin subunit beta 8 |
Gene Aliases: |
- |
Location: |
Chr.7:20329819-20415759 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB8 | NM_002214.2 | 1 | 293 | NP_002205.1 |
| XM_011515393.2 | XP_011513695.1 | |||
| XM_011515394.2 | XP_011513696.1 | |||
| XM_011515396.1 | 1 | 693 | XP_011513698.1 | |
| XM_017012178.1 | XP_016867667.1 | |||
| XM_017012179.1 | XP_016867668.1 | |||
| XM_017012180.1 | 1 | 693 | XP_016867669.1 | |
| XM_017012181.1 | 1 | 693 | XP_016867670.1 | |
| XM_017012183.1 | XP_016867672.1 | |||
| AK295044.1 | 1 | 693 | ||
| BC002630.2 | 1 | 293 | AAH02630.1 | |
| M73780.1 | 1 | 272 | AAA36034.1 |
Target Gene Details
Entrez Gene ID: | 101927811 |
Gene Name: | uncharacterized LOC101927811 |
Gene Aliases: |
- |
Location: |
Chr.7:20328301-20331762 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927811 | NR_110119.1 | |||
| NR_110120.1 | ||||
| AV707592.1 | ||||
| HY150107.1 |
More Information
Set Membership: |
|

Genomic Map