Assay Details
Target Gene Details
Entrez Gene ID: | 51564 |
Gene Name: | histone deacetylase 7 |
Gene Aliases: |
HD7, HD7A, HDAC7A |
Location: |
Chr.12:47782710-47820612 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC7 | NM_001098416.3 | 19 | 2373 | NP_001091886.1 |
| NM_001308090.1 | 19 | 2433 | NP_001295019.1 | |
| NM_015401.4 | 20 | 2484 | NP_056216.2 | |
| XM_011538478.1 | 20 | 2436 | XP_011536780.1 | |
| XM_011538479.1 | 21 | 2878 | XP_011536781.1 | |
| XM_011538480.1 | 19 | 2325 | XP_011536782.1 | |
| XM_011538481.1 | 20 | 2329 | XP_011536783.1 | |
| XM_011538482.1 | 20 | 2413 | XP_011536784.1 | |
| XM_017019455.1 | 19 | 2385 | XP_016874944.1 | |
| AF239243.1 | 17 | 2276 | AAF63491.1 | |
| AK001032.1 | 4 | 224 | BAA91474.1 | |
| AK001190.1 | 12 | 1406 | BAA91545.1 | |
| AK024469.1 | 15 | 2464 | BAB15759.1 | |
| AK027781.1 | 2 | 123 | BAB55363.1 | |
| AK122588.1 | 6 | 3443 | BAC56929.1 | |
| AK128383.1 | 12 | 1327 | ||
| AK225910.1 | ||||
| AK299292.1 | 20 | 2399 | ||
| AK301545.1 | 20 | 2347 | ||
| AK303481.1 | 19 | 2339 | ||
| AK309547.1 | 1 | 59 | ||
| AL117455.1 | 16 | 1951 | CAB55935.1 | |
| AY302468.1 | 20 | 2289 | AAQ18232.1 | |
| AY321367.1 | 19 | 2258 | AAP84704.1 | |
| BC006453.1 | 9 | 908 | ||
| BC020505.2 | 12 | 1451 | AAH20505.2 | |
| BC064840.1 | 19 | 2267 | AAH64840.1 | |
| BT009771.1 | 10 | 1158 | AAP88773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv94n21 | Chr.12:47725779 - 47821397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map