Assay Details
Target Gene Details
Entrez Gene ID: | 3561 |
Gene Name: | interleukin 2 receptor subunit gamma |
Gene Aliases: |
CD132, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1 |
Location: |
Chr.X:71107404-71111631 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL2RG | NM_000206.2 | NP_000197.1 | ||
| AB102793.1 | BAD89384.1 | |||
| AB102794.1 | BAD89385.1 | |||
| AB102795.1 | BAD89386.1 | |||
| AB102797.1 | BAD89388.1 | |||
| AK298656.1 | ||||
| AK314932.1 | ||||
| BC014972.1 | AAH14972.1 | |||
| D11086.1 | BAA01857.1 | |||
| DC345219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map