Assay Details
Target Gene Details
Entrez Gene ID: | 632 |
Gene Name: | bone gamma-carboxyglutamate protein |
Gene Aliases: |
BGP, OC, OCN |
Location: |
Chr.1:156241962-156243332 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BGLAP | NM_199173.5 | 1 | 264 | NP_954642.1 |
| BC113432.1 | 1 | 66 | ||
| BC113434.1 | 1 | 66 | ||
| BU622466.1 | 4 | 481 | ||
| CA428336.1 | ||||
| X51699.1 | CAA35996.1 | |||
| X53698.1 | CAA37736.1 |
Target Gene Details
Entrez Gene ID: | 100527963 |
Gene Name: | PMF1-BGLAP readthrough |
Gene Aliases: |
PMF1 |
Location: |
Chr.1:156212988-156243332 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMF1-BGLAP | NM_001199661.1 | NP_001186590.1 | ||
| NM_001199662.1 | NP_001186591.1 | |||
| NM_001199663.1 | NP_001186592.1 | |||
| NM_001199664.1 | NP_001186593.1 | |||
| CD049192.1 | ||||
| CD388852.1 | ||||
| FY213424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509524 | Chr.1:156238274 - 156308437 on Build GRCh38 | Insertion |
|
| nsv547967 | Chr.1:156241685 - 156248064 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map