Assay Details
Target Gene Details
Entrez Gene ID: | 2021 |
Gene Name: | endonuclease G |
Gene Aliases: |
- |
Location: |
Chr.9:128818402-128822676 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENDOG | NM_004435.2 | 1 | 534 | NP_004426.2 |
| XM_011518347.2 | 1 | 632 | XP_011516649.1 | |
| AK130973.1 | ||||
| BC004922.2 | 1 | 409 | AAH04922.1 | |
| BC016351.1 | 1 | 564 | AAH16351.1 | |
| X79444.1 | 1 | 515 | CAA55963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615412 | Chr.9:128754559 - 128824961 on Build GRCh38 | Loss |
|
| nsv521120 | Chr.9:128803275 - 128829871 on Build GRCh38 | Loss |
|
| nsv615411 | Chr.9:128727172 - 128829871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map