Assay Details
Target Gene Details
Entrez Gene ID: | 129049 |
Gene Name: | small G protein signaling modulator 1 |
Gene Aliases: |
RUTBC2 |
Location: |
Chr.22:24806221-24926846 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGSM1 | NM_001039948.3 | NP_001035037.1 | ||
| NM_001098497.2 | NP_001091967.1 | |||
| NM_001098498.2 | NP_001091968.1 | |||
| NM_133454.3 | NP_597711.1 | |||
| AB275761.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588745 | Chr.22:24836098 - 24888862 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map