Assay Details
Target Gene Details
Entrez Gene ID: | 9989 |
Gene Name: | protein phosphatase 4 regulatory subunit 1 |
Gene Aliases: |
MEG1, PP4(Rmeg), PP4R1 |
Location: |
Chr.18:9546791-9615274 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP4R1 | NM_001042388.2 | 20 | 3118 | NP_001035847.1 |
| NM_005134.3 | 20 | 3067 | NP_005125.1 | |
| NR_052003.1 | 20 | 3085 | ||
| XM_011525775.2 | 20 | 3244 | XP_011524077.1 | |
| XM_011525776.1 | 20 | 3108 | XP_011524078.1 | |
| XM_011525779.2 | 17 | 2911 | XP_011524081.1 | |
| XM_017026105.1 | 19 | 3085 | XP_016881594.1 | |
| XM_017026106.1 | 19 | 2922 | XP_016881595.1 | |
| XM_017026107.1 | 19 | 2937 | XP_016881596.1 | |
| AF100744.1 | 9 | 1285 | AAD43008.1 | |
| AF111106.1 | 20 | 3037 | AAD09818.1 | |
| AF200478.1 | 20 | 3017 | AAL78298.1 | |
| AK025509.1 | 12 | 2225 | ||
| AK027064.1 | 20 | 3069 | ||
| AK292538.1 | 20 | 2998 | ||
| BC060829.1 | 20 | 3019 | AAH60829.1 | |
| U79267.1 | 4 | 664 | AAB50211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833585 | Chr.18:9401632 - 9597293 on Build GRCh38 | Gain+Loss |
|
| nsv1056407 | Chr.18:9144347 - 9553766 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map