Assay Details
Target Gene Details
Entrez Gene ID: | 53833 |
Gene Name: | interleukin 20 receptor subunit beta |
Gene Aliases: |
DIRS1, FNDC6, IL-20R2 |
Location: |
Chr.3:136957865-137011085 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL20RB | NM_144717.3 | 1 | 132 | NP_653318.2 |
| AK296832.1 | ||||
| AK316084.1 | ||||
| AY358305.1 | 1 | 132 | AAQ88672.1 | |
| BC033292.1 | AAH33292.1 | |||
| BC063696.1 | AAH63696.2 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map