Assay Details
Target Gene Details
Entrez Gene ID: | 116986 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 |
Gene Aliases: |
CENTG1, GGAP2, PIKE |
Location: |
Chr.12:57723765-57742201 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGAP2 | NM_001122772.2 | NP_001116244.1 | ||
| NM_014770.3 | NP_055585.1 | |||
| XM_005268625.3 | XP_005268682.1 | |||
| XM_005268626.2 | XP_005268683.1 | |||
| XM_017018772.1 | XP_016874261.1 | |||
| AF384128.1 | AAO39848.1 | |||
| AF413077.1 | AAL04171.1 | |||
| AK122827.1 | ||||
| AK292672.1 | ||||
| AY128689.1 | AAM97540.1 | |||
| BC028020.1 | AAH28020.1 | |||
| D79989.1 |
Target Gene Details
Entrez Gene ID: | 100130776 |
Gene Name: | AGAP2 antisense RNA 1 |
Gene Aliases: |
PUNISHER |
Location: |
Chr.12:57726240-57728356 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGAP2-AS1 | NR_027032.1 | 2 | 575 | |
| AK301303.1 | 2 | 549 | ||
| CN313469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470296 | Chr.12:57718406 - 57783509 on Build GRCh38 | Loss |
|
| nsv826383 | Chr.12:57724297 - 57729864 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map