Assay Details
Target Gene Details
Entrez Gene ID: | 8382 |
Gene Name: | NME/NM23 family member 5 |
Gene Aliases: |
NM23-H5, NM23H5, RSPH23 |
Location: |
Chr.5:138115172-138139443 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME5 | NM_003551.2 | 3 | 322 | NP_003542.1 |
| XM_005272099.1 | 2 | 249 | XP_005272156.1 | |
| XM_006714716.3 | 2 | 249 | XP_006714779.1 | |
| XM_017009945.1 | 2 | 249 | XP_016865434.1 | |
| AF067724.1 | 3 | 292 | AAC64358.1 | |
| AK312181.1 | 3 | 269 | ||
| BC026182.2 | 3 | 294 | AAH26182.1 | |
| KU178416.1 | 2 | 244 | ||
| KU178417.1 | ||||
| U90450.1 | 3 | 268 | AAC69440.1 | |
| Y14992.1 | 2 | 258 | CAA75226.1 |
More Information
Set Membership: |
|

Genomic Map