Assay Details
Target Gene Details
Entrez Gene ID: | 79770 |
Gene Name: | thioredoxin domain containing 15 |
Gene Aliases: |
C5orf14, UNQ335 |
Location: |
Chr.5:134873770-134901633 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNDC15 | NM_024715.3 | 1 | 431 | NP_078991.3 |
| AK056512.1 | 1 | 466 | BAB71199.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830494 | Chr.5:134825197 - 134984298 on Build GRCh38 | Gain+Loss |
|
| nsv830493 | Chr.5:134667777 - 134926221 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map