Assay Details
Target Gene Details
Entrez Gene ID: | 3718 |
Gene Name: | Janus kinase 3 |
Gene Aliases: |
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK |
Location: |
Chr.19:17824782-17848071 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAK3 | NM_000215.3 | NP_000206.2 | ||
| XM_005259896.3 | XP_005259953.1 | |||
| XM_011527990.1 | XP_011526292.1 | |||
| XM_011527991.2 | XP_011526293.1 | |||
| AK296383.1 | ||||
| BC028068.1 | AAH28068.1 | |||
| KU178153.1 | ||||
| KU178154.1 | ||||
| U09607.1 | AAA19626.1 | |||
| U31601.1 | AAC50226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv50n68 | Chr.19:17671882 - 17842923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map