Assay Details
Target Gene Details
Entrez Gene ID: | 10112 |
Gene Name: | kinesin family member 20A |
Gene Aliases: |
MKLP2, RAB6KIFL |
Location: |
Chr.5:138178728-138187715 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF20A | NM_005733.2 | NP_005724.1 | ||
| AF070672.1 | AAC83230.1 | |||
| AF153329.1 | AAD37806.1 | |||
| AK025790.1 | ||||
| AK296879.1 | ||||
| BC012999.2 | AAH12999.1 |
More Information
Set Membership: |
|

Genomic Map