Assay Details
Target Gene Details
Entrez Gene ID: | 10589 |
Gene Name: | DR1 associated protein 1 |
Gene Aliases: |
NC2-alpha |
Location: |
Chr.11:65919257-65921577 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRAP1 | NM_006442.3 | 7 | 864 | NP_006433.2 |
| BC010025.2 | 7 | 672 | AAH10025.1 | |
| BQ773919.1 | 1 | 159 | ||
| X96506.1 | 7 | 619 | CAA65358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
| nsv1159905 | Chr.11:65836608 - 65922038 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map