Assay Details
Target Gene Details
Entrez Gene ID: | 51166 |
Gene Name: | aminoadipate aminotransferase |
Gene Aliases: |
KAT2, KATII, KYAT2 |
Location: |
Chr.4:170060222-170091699 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADAT | NM_001286682.1 | 6 | 745 | NP_001273611.1 |
| NM_001286683.1 | 6 | 818 | NP_001273612.1 | |
| NM_016228.3 | 5 | 951 | NP_057312.1 | |
| NM_182662.1 | 6 | 733 | NP_872603.1 | |
| XM_006714231.2 | 7 | 875 | XP_006714294.1 | |
| XM_011532020.2 | 3 | 508 | XP_011530322.1 | |
| AF481738.1 | 5 | 952 | AAM09683.1 | |
| AK055952.1 | 6 | 733 | ||
| AK314729.1 | 6 | 820 | ||
| BC031068.1 | 6 | 738 | AAH31068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4601 | Chr.4:170048721 - 170093413 on Build GRCh38 | Deletion |
|
| nsv830150 | Chr.4:170072762 - 170230617 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map