Assay Details
Target Gene Details
Entrez Gene ID: | 10912 |
Gene Name: | growth arrest and DNA damage inducible gamma |
Gene Aliases: |
CR6, DDIT2, GADD45gamma, GRP17 |
Location: |
Chr.9:89605009-89606555 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GADD45G | NM_006705.3 | 3 | 353 | NP_006696.1 |
| XM_011518163.2 | 2 | 254 | XP_011516465.1 | |
| AF078078.1 | 3 | 258 | AAC83329.1 | |
| AF079806.1 | 3 | 340 | AAD28544.1 | |
| AF087883.1 | 3 | 338 | AAK00414.1 | |
| AK313689.1 | 3 | 351 | ||
| BC000465.2 | 3 | 334 | AAH00465.1 | |
| BC019325.1 | 3 | 334 | AAH19325.1 | |
| BT007234.1 | 3 | 243 | AAP35898.1 | |
| CR457038.1 | 3 | 243 | CAG33319.1 | |
| D83023.1 | 3 | 326 | BAA84543.1 | |
| DA420966.1 | 3 | 353 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain |
|
| nsv1047460 | Chr.9:89512864 - 89727875 on Build GRCh38 | Gain |
|
| nsv509309 | Chr.9:89584439 - 89677102 on Build GRCh38 | Insertion |
|
| esv29434 | Chr.9:89348488 - 89728467 on Build GRCh38 | Gain+Loss |
|
| nsv469563 | Chr.9:89545246 - 89723819 on Build GRCh38 | Gain |
|
| nsv471662 | Chr.9:89545247 - 89723819 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map