Assay Details
Target Gene Details
Entrez Gene ID: | 1725 |
Gene Name: | deoxyhypusine synthase |
Gene Aliases: |
DHS, DS, MIG13 |
Location: |
Chr.19:12672164-12681887 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHPS | NM_001206974.1 | NP_001193903.1 | ||
| NM_001930.3 | NP_001921.1 | |||
| NM_013406.2 | NP_037538.1 | |||
| NR_038192.1 | ||||
| XM_011527770.1 | XP_011526072.1 | |||
| XM_011527771.2 | XP_011526073.1 | |||
| AK291553.1 | ||||
| AK300498.1 | ||||
| AK304780.1 | ||||
| AL520040.3 | ||||
| AY336090.1 | AAR24620.1 | |||
| BC000333.2 | AAH00333.1 | |||
| BC014016.2 | AAH14016.1 | |||
| L39068.1 | AAA86282.1 | |||
| U26266.1 | AAB02175.1 | |||
| U32178.1 | AAB02179.1 | |||
| U40579.1 | AAA96151.1 | |||
| U79262.1 | AAB50208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication |
|
| dgv138n111 | Chr.19:12627853 - 12810372 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map